Learn More
BrandTech™ VITALAB™ Polypropylene Sample Vials with Snap-On Caps
These Polypropylene Sample Vials with Snap-On Caps are perfectly suited for the collection, storage and transport of samples in the laboratory.
Brand:  BrandTech™ 130594
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10360275
  • £92.20 / Pack of 10
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Transparent in color
  • Snap caps and vial body from PP
  • Excellent resistance to alcohols, aldehydes, alkalis and acids
Specifications
Specifications
Transparent
Polypropylene
10
Polypropylene
112 mm
GL 56
54 mm
180 mL
125°C Max.
Screw Cap
Sample Container