Learn More
Erlab Transparent Back Panel Captair 481 Smart
Click to view available options
For Use With (Equipment):
Fuming Chamber
Unit Size:
Each
Description
- Transparent back panel for your ductless fume hood Captair 481 Smart
- Material: Tempered glass
Specifications
Specifications
| For Use With (Equipment) | Fuming Chamber |
| Product Type | Back Panel |