missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap

Accurately measure precise volumes of a liquid. Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap is used for dilutions and to prepare chemical solutions.

Brand:  Kartell™ 0143400

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10587051

  • £124.00 / Pack of 10
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

Accurately measure precise volumes of a liquid. Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap is used for dilutions and to prepare chemical solutions.

These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Specifications

Specifications

Translucent
Polypropylene
Screw Caps
10
Autoclavable
500 mL
265 mm
GL25
±5 mL
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers