Learn More
Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap
Brand:  Kartell™ 0143300
Product Code. 10776912
  • ONLINE EXCLUSIVE PRICE
    £116.00
    £104.55 / Pack of 10
    Save £11.45
    9% Off
Only null left
Estimated Shipment: 30-01-2026
Add to basket
Description
Description
These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.