missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Translin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | Translin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Translin Polyclonal specifically detects Translin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Translin | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_004613 | |
| 7247 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human Translin. The immunogen is located within the last 50 amino acids of Translin. Amino Acid Squence: NLKNDSLRKRYDGLKYDVKK | |
| Primary | |
| Store at 4°C. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| BCLF-1, RCHF1, recombination hotspot associated factor, recombination hotspot-binding protein, REHF-1, TBRBP, translin, TRSLN | |
| TSN | |
| IgG | |
| Affinity Purified | |
| At least two isoforms of Translin are known to exist; this antibody will detect both. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title