missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat Apoptosis regulator Bcl-2 siRNA
Blocks transcription and translation of Apoptosis regulator Bcl-2 from Rat.
£227.54 - £336.64
Specifications
Accession Number | NP_058689 |
---|---|
Cell Lines Tested | Pc12 Pheochromocytoma Cells of the Rat Adrenal Gland (Medulla). |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Bcl2 |
Gene ID (Entrez) | 24224 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099161
|
enQuireBio™
QX11-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089161
|
enQuireBio™
QX11-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058689 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
24224 | |
Bcl-2, PPP1R50 | |
Lyophilized |
Pc12 Pheochromocytoma Cells of the Rat Adrenal Gland (Medulla). | |
Bcl2 | |
siRNA | |
Rat |