missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transketolase Antibody (OTI5H3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74585CL1
This item is not returnable.
View return policy
Description
Transketolase Monoclonal antibody specifically detects Transketolase in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Transketolase | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase | |
| Full length human recombinant protein of human TKT (NP_001055) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 7086 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI5H3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction