missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Transketolase Polyclonal specifically detects Transketolase in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Transketolase |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Transketolase. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?