missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 7/TGM7 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81763AF350
This item is not returnable.
View return policy
Description
Transglutaminase 7/TGM7 Polyclonal antibody specifically detects Transglutaminase 7/TGM7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Transglutaminase 7/TGM7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 350 | |
| protein-glutamine gamma-glutamyltransferase Z, TG(Z), TGase Z, TGase-7, TGMZEC 2.3.2.13, TGZ, transglutaminase 7, Transglutaminase Z, transglutaminase-7 | |
| TGM7 antibody was raised against a 13 amino acid synthetic peptide near the center of human TGM7. The immunogen is located within amino acids 460 - 510 of TGM7. Amino Acid Squence: LESGGLRDQPAQL | |
| 0.1 mL | |
| Cancer | |
| 116179 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction