missing translation for 'onlineSavingsMsg'
Learn More

Transglutaminase 5 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18428446 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428446 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428446 Supplier Novus Biologicals Supplier No. NBP281857FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Transglutaminase 5 Polyclonal antibody specifically detects Transglutaminase 5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transglutaminase 5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.2.13, protein-glutamine gamma-glutamyltransferase 5, TG(X), TGase X, TGase-5, TGASE5, TGASEX, TGM6, TGMX, TGXMGC141907, transglutaminase 5, transglutaminase V, Transglutaminase X, transglutaminase-5
Host Species Rabbit
Immunogen Transglutaminase 5 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human Transglutaminase 5. The immunogen is located within the last 50 amino acids of Transglutaminase 5. Amino Acid Squence: NMRSNKFKDIKGYRNVYVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9333
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.