missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 5 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81857AF488
This item is not returnable.
View return policy
Description
Transglutaminase 5 Polyclonal antibody specifically detects Transglutaminase 5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Transglutaminase 5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| EC 2.3.2.13, protein-glutamine gamma-glutamyltransferase 5, TG(X), TGase X, TGase-5, TGASE5, TGASEX, TGM6, TGMX, TGXMGC141907, transglutaminase 5, transglutaminase V, Transglutaminase X, transglutaminase-5 | |
| Transglutaminase 5 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human Transglutaminase 5. The immunogen is located within the last 50 amino acids of Transglutaminase 5. Amino Acid Squence: NMRSNKFKDIKGYRNVYVD | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 9333 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction