missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Transglutaminase 2/TGM2 Antibody (zampilimab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Transglutaminase 2/TGM2 |
|---|---|
| Clone | zampilimab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30048581
|
Novus Biologicals™
NBP3-28563-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046392
|
Novus Biologicals™
NBP3-28563-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046030
|
Novus Biologicals™
NBP3-28563-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transglutaminase 2/TGM2 Monoclonal antibody specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Transglutaminase 2/TGM2 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
| TGM2/ Transglutaminase 2 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| zampilimab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P21980 | |
| 7052 | |
| IgG4 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title