missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 2/TGM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£183.00 - £341.00
Specifications
| Antigen | Transglutaminase 2/TGM2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-5 ug/ml, Simple Western 1:50, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18129321
|
Novus Biologicals
NBP2-24470SS |
0.025 mg |
£183.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18741274
|
Novus Biologicals
NBP2-24470 |
0.1 mg |
£341.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transglutaminase 2/TGM2 Polyclonal specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Transglutaminase 2/TGM2 | |
| Western Blot 2-5 ug/ml, Simple Western 1:50, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
| TGM2 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, 93% homologous in chimpanzee and monkey. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P21980 | |
| 7052 | |
| A portion of amino acids 600-650 of human Transglutaminase 2 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title