missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ TransFix™ Cellular Antigen Stabilizing Reagent

Produktkod. 10465503
missing translation for 'orderingAttributeHoverText'
Quantity:
20 mL
Förpackningsstorlek
Each
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 10465503

missing translation for 'mfr': Invitrogen™ FIX20

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

TransFix™ Cellular Antigen Stabilizing Reagent

Introducing TransFix™ Reagent - a whole blood preservative that stabilizes leukocytes and preserves antigenic sites for future cellular analysis (Figure 1). Now it's simple to batch-process multiple samples in order to optimize laboratory workflow. Additionally, blood that has been stabilized with TransFix™ Reagent can be collected and transported to other sites for analysis. TransFix™ Reagent is:

  • Easy to use-just add to whole blood, mix, and store for up to 10 days at 4°C
  • Cost effective-batch samples to optimize laboratory work-flow while maintaining sample integrity
  • Proven-multiple publications demonstrate its effectiveness at preserving a broad range of cellular antigens (Table 1)
  • Convenient-compatible with EDTA (K3), ACD, and heparin anticoagulants
  • Flexible-preserves a wide range of samples, including human, mouse, rat, sheep, pig, and more

    View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.

    References
    1. Barnett, D. et al. (1999) Cytometry 38:88;
    2. Jani, I.V. et al. (2001) J Immunol Methods 257:145;
    3. Bergeron, M. et al. (2002) Cytometry 50:86.

    TransFix™ Reagent is a proprietary whole blood stabilization solution, and is a general purpose reagent (GPR).
    TransFix™ is a registered trademark of Sheffield Teaching Hospitals NHS Trust.
  • TRUSTED_SUSTAINABILITY

    Specifikationer

    Description TransFix™ Cellular Antigen Stabilizing Reagent, 20mL, Room Temperature Shipping Condition, 18 to 25°C Storage Requirements, Convenient-Compatible with EDTA (K3), ACD and Heparin Anticoagulants For Flow Cytometry
    Product Type TransFix™ Cellular Antigen Stabilizing Reagent
    Quantity 20 mL
    For Use With (Application) Flow Cytometry
    Content And Storage 18°C to 25°C
    Packaging Quantity 1 Each

    For Research Use Only. Not for use in diagnostic procedures.

    Korrigering av produktinnehåll

    Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

    Produkttitel

    Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

    Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.