Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (T56/14), PE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MHCD7104-4
 View more versions of this product
Product Code. 10343733
  • £784.00 / 2mL
For Research Use Only. All usage must comply with product instructions.
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transferrin Receptor, CD71
Monoclonal
PE
TFRC
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr
Mouse
Purified
ASR
7037
4°C, store in dark
Liquid
Flow Cytometry
T56/14
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P02786
TFRC
Human CD71.
2 mL
Primary
Human
Antibody
IgG1