missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (OX-26)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517555
This item is not returnable.
View return policy
Description
Transferrin Receptor Monoclonal Antibody for Western Blot, IHC (F), Flow, IP
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
Specifications
| Transferrin Receptor | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q99376 | |
| TFRC | |
| PHA activated lymph node cells of a PVG rat. | |
| 500 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| OX-26 | |
| Unconjugated | |
| TFRC | |
| 2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
| Mouse | |
| Protein G | |
| RUO | |
| 64678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction