missing translation for 'onlineSavingsMsg'
Learn More

Transferrin Receptor Monoclonal Antibody (H68.4), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen 136890

Code : 50

Additional Details : Weight : 0.50000kg

Product Code. 10516083

  • £834.00 / Each
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

H68.4 is specific to residues 3-28 of the human transferrin receptor (TfR) tail.

Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
Specifications

Specifications

Transferrin Receptor
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide; pH 7.4
P02786, Q07891, Q62351, Q90997, Q99376
TFRC
Recombinant human transferrin receptor.
500 μg
Primary
Chicken, Hamster, Human, Mouse, Rat
Antibody
IgG1
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot
H68.4
Unconjugated
TFRC
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr
Mouse
Affinity chromatography
RUO
100689395, 22042, 396191, 64678, 7037
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers