missing translation for 'onlineSavingsMsg'
Learn More

Transferrin, Rabbit anti-Human, Clone: TW4.20, Novus Biologicals™

Product Code. 18384991 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384991 0.025 mg 25µg
18388970 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18384991 Supplier Novus Biologicals Supplier No. NBP2810940.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transferrin
Applications Western Blot, Flow Cytometry, ELISA, Blocking Assay
Classification Monoclonal
Clone TW4.20
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Block/Neutralize
Formulation PBS
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene Symbols TF
Host Species Rabbit
Immunogen Iron-free purified human transferrin.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity Binds to the N-terminal domain of human transferrin.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.