missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin R (CD71) Mouse anti-Human , PE, Clone: OKT9 (also known as OKT-9; OKT 9), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566722
This item is not returnable.
View return policy
Description
Transferrin R (CD71) Monoclonal antibody specifically detects Transferrin R (CD71) in Human samples. It is validated for Flow Cytometry
Specifications
| Transferrin R (CD71) | |
| Monoclonal | |
| PE | |
| TfR; sTfR; TFRC; Trfr; TfR1; TR; T9; p90 | |
| Human T-cell acute lymphoblastic leukemia cells | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| OKT9 (also known as OKT-9; OKT 9) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction