missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Transferrin Native Protein
Shop All Bio Techne Products
Click to view available options
:
1mg; Biotin
Description
Native Transferrin. Source: Serum The Transferrin Native Protein has been validated for the following applications: Western Blot, ELISA.
Specifications
Specifications
| Formulation | Lyophilized from 0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free |
| Gene ID (Entrez) | 7018 |
| Name | Transferrin Protein |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction