missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Transferrin Native Protein

Product Code. 18074627 Shop All Bio Techne Products
Change view
Click to view available options
:
1mg; Rhodamine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18074627 1mg; Rhodamine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18074627 Supplier Novus Biologicals™ Supplier No. NBP196857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

Native Transferrin. Source: Serum The Transferrin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q06AH7
For Use With (Application) Dot Blot
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000)
Gene ID (Entrez) 7018
Name Transferrin Protein
Storage Requirements 4°C in the dark.
Cross Reactivity Human
Purity or Quality Grade Multi-step
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.