missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin, Mouse anti-Human, Clone: TF/3001, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79778-100UG
This item is not returnable.
View return policy
Description
Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Transferrin | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P02787 | |
| TF | |
| Recombinant fragment (around aa 311-445) of human Transferrin protein (exact sequence is proprietary) | |
| 100 μg | |
| Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers | |
| 7018 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| TF/3001 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction