missing translation for 'onlineSavingsMsg'
Learn More

Transferrin Mouse anti-Human, Clone: 2B4C6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66171-1-IG-150UL

Product Code. 16827805

  • £189.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis. Serum transferrin may also have a further role in stimulating cell proliferation.
Specifications

Specifications

Transferrin
Monoclonal
0.89 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02787
TF
Transferrin Fusion Protein Ag11668
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2B4C6
Unconjugated
TF
Beta 1 metal binding globulin, PRO1557, PRO2086, Serotransferrin, Siderophilin, TF, Transferrin
Mouse
Protein G
RUO
7018
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.