missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Transferrin, HRP

Product Code. 10240064
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10240064 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10240064 Supplier Invitrogen™ Supplier No. 31453
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product Purity: The precipitin line(s) against goat anti-mouse whole serum is (are) the same as that (those) against goat anti-mouse transferrin when assayed by immunoelectrophoresis at an antigen concentration of 20 mg/mL. Transferrin Concentration: 1.0 mg/mL after reconstitution. Restoration and Storage: Store product at 4°C until opened. Restore with 1.1 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. (To judge clarity, draw product into a clean pasteur pipette). Product is stable for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of 50% glycerol and store at -20°C. Country of Origin: USA.

Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Lyophilized
Product Type Transferrin
Quantity 1 mg
Concentration 1 mg/mL
Formulation PBS with 15mg/mL BSA and no preservative; pH 7.6
For Use With (Application) Control, Immunohistochemistry, Blocking Assay
Isotype Ig
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.