man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Recombinant Proteins
Fisher BioReagents™ Transferrin, Guinea Pig
Print
Fisher BioReagents™ Transferrin, Guinea Pig
£311.00
See promotional offers below
Specifications
Packaging
Amber Glass
View More Specs
Products
1
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
1
Product Code
Brand
Quantity
Price
Quantity & Availability
Product Code
Brand
Quantity
Price
Quantity & Availability
10411545
SDS
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Fisher BioReagents™
BP2544-10
10mg
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
View alternative products
Save Up to
»
Description
Description
Specifications
Specifications
Packaging
Amber Glass
Videos
SDS
Documents
Documents
Product Certifications
Back to Top