missing translation for 'onlineSavingsMsg'
Learn More

Transferrin Antibody, Novus Biologicals™

Product Code. 18351835 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351835 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351835 Supplier Novus Biologicals Supplier No. NBP314692

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Transferrin Polyclonal antibody specifically detects Transferrin in Equine samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Transferrin
Applications Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with 0.09% Sodium Azide
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Host Species Rabbit
Immunogen no immunogen data
Purification Method Immunoelectrophoresis (IEP)
Quantity 2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Equine
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.