missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ Transferpette™ pro Shelf/Rack Mounts

Product Code. 30696131 Shop All Brand Products
Change view
Click to view available options
Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
30696131 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30696131 Supplier BRANDTECH™ Supplier No. 704881
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Spare part for Transferpette™ pro

  • Spare part
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color Light Gray
Holds Manual Pipette
Material Polycarbonate
Positions 1
Shape Z-shaped
Stackable No
Sterility Non-sterile
For Use With (Equipment) Transferpette pro
Product Type Shelf Pipette Holder
Shelves Removable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.