Description
- Spare part
Specifications
Specifications
| For Use With | Transferpette Pro |
| Material | FKM, Steel |
| Type | Seal with Spring |
| For Use With (Equipment) | Transferpette pro 30 to 300 μL |