Learn More
BRANDTECH™ Transferpette™ pro Piston Shaft Unit
Product Code. 30696134 Shop All Brand Products
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
30696134 Each
1 options
Product Code. 30696134 Supplier BRANDTECH™ Supplier No. 705675
Only null left
Add to basket
Add to basket
Spare part for Transferpette™ pro
  • Spare part
TRUSTED_SUSTAINABILITY
Specifications
For Use With Transferpette Pro
Material PEEK, PEI
Type Piston Shaft
For Use With (Equipment) Transferpette pro 50 to 1250 μL