missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Transferpette™ -12 Electronic Microliter Pipette
Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.
Brand: Brand™ 705464
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Functional, ergonomic housing design
- 12 channels
- Individually adjustable finger rest
- Intuitive menu structure and comprehensively illustrated user manual
- Significantly reduced tip attachment and ejection forces using universal tip
- Corrosion-resistant piston and ejector
- Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis, dispensing
- Ready for use: 4000 pipetting cycles with each battery charge, battery refresh function even during recharging
- Each pipette has a DE-M marking and is supplied with performance certificate
- Supplied with battery, AC adapter, device stand, reagent reservoir, mounting tool, silicone oil and one set of sealing rings made of FKM
Specifications
≤ 0.8%, ± 1.6μL | |
Autoclavable | |
12 | |
Yes | |
10 to 200 μL | |
Ergonomic | |
0.2μL | |
230V/50 Hz | |
50 Hz |
360° | |
Yellow | |
Transferpette™ | |
Electronic Microliter Pipette | |
LCD | |
UK/Ireland AC Adapter | |
1 Each | |
230 V |