missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Transfer Rack for Eppendorf SmartBlock™

Transfer rack suitable for individually sensor-controlled SmartBlock™ heating/cooling/mixing blocks.

£44.41

Specifications

For Use With (Equipment) SmartBlock, ThermoMixer
Temperature (Metric) Range −86°C to 121 °C
View More Specs

Products 2
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
15541290
missing translation for 'viewDocuments' Promotion Details
Eppendorf™
3880000151
24 x 1.5/2.0 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
15551290
missing translation for 'viewDocuments' Promotion Details
Eppendorf™
3880000305
24 x 0.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

  • Holds up to 24 microtubes for easy transfer of multiple vessels in parallel.
  • Convenient transfer of up to 24 hot vessels without burning the fingers
  • Temperature range from -86°C to 121°C for broad flexibility
  • Limited to ThermoMixer C annd ThermoStat C
Specifications

Specifications

SmartBlock, ThermoMixer
−86°C to 121 °C
Videos
SDS
Documents

Documents

Certificates