Learn More
Thermo Scientific Chemicals Transfer or electro blotting buffer (10X), pH 8.5
Description
Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Form | Liquid |
| Quantity | 4L |
| Storage Requirements | Ambient temperatures |
Frequently Asked Questions (FAQs)
You can make a 1:10 dilution of 10X Transfer or electro blotting buffer (10X), pH 8.5 with deionized water to create a 1X Transfer or electro blotting buffer. Adjust the pH to 8.5.
Transfer or electro blotting buffer (10X), pH 8.5 consists of 250 mM Tris and 1.92 M glycine.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.