missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Transfer or electro blotting buffer (10X), pH 8.5

Product Code. 15455309
Click to view available options
Quantity:
1L
2L
4L
Unit Size:
1 L
2 L
4 L
This item is not returnable. View return policy

Product Code. 15455309

Brand: Thermo Scientific Chemicals J63037.K3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.

Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 2L
Storage Requirements Ambient temperatures
How can I prepare a 1X buffer solution from the 10X Transfer or electro blotting buffer (10X), pH 8.5?

You can make a 1:10 dilution of 10X Transfer or electro blotting buffer (10X), pH 8.5 with deionized water to create a 1X Transfer or electro blotting buffer. Adjust the pH to 8.5.

What are the concentrations of the components in the Transfer or electro blotting buffer (10X), pH 8.5?

Transfer or electro blotting buffer (10X), pH 8.5 consists of 250 mM Tris and 1.92 M glycine.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.