missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Transfer/Electro Blotting 10X Solution
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Formulation:
TRIS Base: 0.25M
Glycine: 1.92M
pH: 8.5
Add Methanol according to protocol
Quantity: 1LSpecifications
Specifications
| CAS | 77-86-1 |
| Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction