missing translation for 'onlineSavingsMsg'
Learn More

Transcrocetinate disodium, MedChemExpress

Product Code. 16417068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417068

Brand: Medchem Express HY16502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Transcrocetinate disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity.

Transcrocetinate disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity.

Specifications

Chemical Name or Material Transcrocetinate disodium
Physical Form Solid
CAS 591230-99-8
Color Reddish Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H22Na2O4
For Use With (Application) Cancer-programmed cell death
Synonym Disodium trans-crocetinate
Solubility Information H2O : 16.67 mg/mL (44.77 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(O[Na])/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C(O[Na])=O
Molecular Weight (g/mol) 372.37
Formula Weight 372.37
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.