missing translation for 'onlineSavingsMsg'
Learn More

Transcrocetin, MedChemExpress

Product Code. p-200101241
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16439258

Brand: Medchem Express HYN2072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Transcrocetin (trans-Crocetin), extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. Transcrocetin (trans-Crocetin) is capable of crossing the blood-brain barrier and reach the central nervous system (CNS).

Transcrocetin (trans-Crocetin), extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. Transcrocetin (trans-Crocetin) is capable of crossing the blood-brain barrier and reach the central nervous system (CNS).

Specifications

Chemical Name or Material Transcrocetin
CAS 27876-94-4
Color Orange
Purity Grade Notes Research
Molecular Formula C20H24O4
Quantity 10 mg
Synonym trans-Crocetin
Solubility Information DMSO : 1 mg/mL (3.05 mM; Need ultrasonic and warming)
SMILES O=C(O)/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C(O)=O
Molecular Weight (g/mol) 328.4
Formula Weight 328.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.