missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Transcription Factor Phospho Buffer Set (RUO)

Product Code. p-7162080
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15844409

Brand: BD Biosciences™ 565575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed to enable robust and simultaneous flow cytometric analysis of various cell surface antigens, transcription factors, and phosphorylated signal proteins in cells derived from both human and mouse tissues.

  • Designed to enable robust and simultaneous flow cytometric analysis of various cell surface antigens, transcription factors, and phosphorylated signal proteins in cells derived from both human and mouse tissues.
  • This includes fresh, fixed, and frozen human peripheral blood mononuclear cells.
  • The BD Pharmingen™ TFP Buffer Set has been successfully applied to the multiparameter flow cytometric analysis of phosphorylated Stat signaling responses to cytokines made by human and mouse T regulatory cell and conventional T cell subsets, and other leucocyte types defined by certain cell surface markers, transcription factors (FoxP3 and T-bet), and physical characteristics.
  • While not all antibody compatibilities are known, cell signaling responses to different stimuli should also be amenable to analysis using this buffer set and antibodies specific for other cell surface markers, transcription factors, and types of downstream phosphorylated signaling proteins.
  • This buffer system is backwards compatible with many BD reagents, including antibodies conjugated with new fluorescent dyes (eg, BD Horizon™ BV421, BV510, PE-CF594, V450, and V500).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Transcription Factor Phospho Buffer Set
Quantity 25 Tests
Formulation 20 mL 0.75X TFP Diluent Buffer, 40 mL 5X TFP Fix/Perm Buffer, 25 mL Perm Buffer III, 7.5 mL 4X TFP Perm/Wash Buffer
Includes 1 Set, 25 Tests, 20 mL 0.75X TFP Diluent Buffer, 40 mL 5X TFP Fix/Perm Buffer, 25 mL Perm Buffer III, 7.5 mL 4X TFP Perm/Wash Buffer
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Transcription Factor Phospho Buffer Set
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : formaldehyde, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.