missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcription factor E3 Antibody (TFE3/6849R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21005C
This item is not returnable.
View return policy
Description
Transcription factor E3 Monoclonal antibody specifically detects Transcription factor E3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Transcription factor E3 | |
| Monoclonal | |
| DyLight 650 | |
| BHLHE33, bHLHe33transcription factor E3, Class E basic helix-loop-helix protein 33, RCCP2, TFEA, transcription factor binding to IGHM enhancer 3, transcription factor E family, member A, Transcription factor for IgH enhancer | |
| Recombinant full-length human Transcription factor E3 protein | |
| 0.1 mL | |
| Cancer, Immunology | |
| 7030 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TFE3/6849R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction