missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcription factor E3 Antibody (CL12810) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18574AF405
This item is not returnable.
View return policy
Description
Transcription factor E3 Monoclonal antibody specifically detects Transcription factor E3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Transcription factor E3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| BHLHE33, bHLHe33transcription factor E3, Class E basic helix-loop-helix protein 33, RCCP2, TFEA, transcription factor binding to IGHM enhancer 3, transcription factor E family, member A, Transcription factor for IgH enhancer | |
| This antibody was developed using a recombinant protein derived from P19532, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Immunology | |
| 7030 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL12810 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction