missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcobalamin II Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98341CL1
This item is not returnable.
View return policy
Description
Transcobalamin II Polyclonal antibody specifically detects Transcobalamin II in Mouse samples. It is validated for ELISA
Specifications
| Transcobalamin II | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| CoraFluor 1 | |
| D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2 | |
| Produced in rabbits immunized with purified, recombinant Mouse Transcobalamin II (Uniprot#: O88968; Met 1-Trp 430) | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 6948 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction