missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcobalamin II Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94355-0.1ml
This item is not returnable.
View return policy
Description
Transcobalamin II Polyclonal antibody specifically detects Transcobalamin II in Human, Mouse samples. It is validated for Western Blot
Specifications
| Transcobalamin II | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-250 of human TCN2 (NP_001171655.1). EMCEIPEMDSHLVEKLGQHLLPWMDRLSLEHLNPSIYVGLRLSSLQAGTKEDLYLHSLKLGYQQCLLGSAFSEDDGDCQGKPSMGQLALYLLALRANWHDHKGHPHTSYYQYGLGILALCLHQKRVHDSVVDKLLYAVEPFHQGHHSVDTAAMAGLAFTCLKRSNFNPGRRQRITMAIRTVREEILKAQTPEGHFGNVYSTPLALQFLMTSPMRGAELGTACLKARVALLAS | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 6948 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido