missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcobalamin II Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94355-0.02ml
This item is not returnable.
View return policy
Description
Transcobalamin II Polyclonal antibody specifically detects Transcobalamin II in Human, Mouse samples. It is validated for Western Blot
Specifications
| Transcobalamin II | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-250 of human TCN2 (NP_001171655.1). EMCEIPEMDSHLVEKLGQHLLPWMDRLSLEHLNPSIYVGLRLSSLQAGTKEDLYLHSLKLGYQQCLLGSAFSEDDGDCQGKPSMGQLALYLLALRANWHDHKGHPHTSYYQYGLGILALCLHQKRVHDSVVDKLLYAVEPFHQGHHSVDTAAMAGLAFTCLKRSNFNPGRRQRITMAIRTVREEILKAQTPEGHFGNVYSTPLALQFLMTSPMRGAELGTACLKARVALLAS | |
| 0.02 mL | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 6948 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction