missing translation for 'onlineSavingsMsg'
Learn More

Transcobalamin II Antibody (049), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18432078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432078 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432078 Supplier Novus Biologicals Supplier No. NBP289597JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Transcobalamin II Monoclonal antibody specifically detects Transcobalamin II in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transcobalamin II
Applications ELISA
Classification Monoclonal
Clone 049
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Transcobalamin II (O88968; Met1-Trp430).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6948
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.