missing translation for 'onlineSavingsMsg'
Learn More

Transaldolase 1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-32216JF646

Product Code. 18794842

  • £492.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Transaldolase 1 Polyclonal antibody specifically detects Transaldolase 1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transaldolase 1
Polyclonal
Western Blot
dihydroxyacetone transferase, EC 2.2.1.2, glycerone transferase, TAL, TALDO, TALDOR, TALH, TAL-H, transaldolase, transaldolase 1
The immunogen this antibody was made to, maps to a region between residue 200 to 250 of human Transaldolase 1 using the numbering given in entry NP_006746.1 (GeneID 6888).
0.1 mL
Stem Cell Markers
6888
Store at 4°C in the dark.
IgG
Western Blot
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.