missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transaldolase 1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32217UV
This item is not returnable.
View return policy
Description
Transaldolase 1 Polyclonal antibody specifically detects Transaldolase 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Transaldolase 1 | |
| Polyclonal | |
| Western Blot | |
| dihydroxyacetone transferase, EC 2.2.1.2, glycerone transferase, TAL, TALDO, TALDOR, TALH, TAL-H, transaldolase, transaldolase 1 | |
| The immunogen this antibody was made to, maps to a region between residue 267 to 317 of human Transaldolase 1 using the numbering given in entry NP_006746.1 (GeneID 6888). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 6888 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion