missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin, TRC

Product Code. 30133043
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133043 25 mg 25mg
30133627 50 mg 50mg
30007057 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133043 Supplier Toronto Research Chemicals Supplier No. TRCZ27300025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-39-4
Formula Weight 219.112
InChI Formula InChI=1S/C10H13N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h2,5-6,16H,3-4H2,1H3,(H2,11,12,13,14,15)/b7-2+
IUPAC Name (E)-2-methyl-4-(9H-purin-6-ylamino)but-2-en-1-ol
Molecular Formula C10 H13 N5 O
Molecular Weight (g/mol) 219.24
Percent Purity >95%
SMILES C\C(=C/CNc1ncnc2[nH]cnc12)\CO
Synonym (2E)-2-Methyl-4-(9H-purin-6-ylamino)-2-buten-1-ol (ACI), 2-Buten-1-ol, 2-methyl-4-(1H-purin-6-ylamino)-, (2E)- (9CI), 2-Buten-1-ol, 2-methyl-4-(1H-purin-6-ylamino)-, (E)- (ZCI), 2-Buten-1-ol, 2-methyl-4-(purin-6-ylamino)-, (E)- (8CI), Zeatin (7CI), (E)-Zeatin, 6-(4-Hydroxy-3-methyl-trans-2-butenylamino)purine, MB 907, N6-(4-Hydroxy-3-methyl-trans-2-butenyl)adenine, trans-6-(4-Hydroxy-3-methylbut-2-enyl)amino purine, trans-Zeatin, Zeatine, ZT, ZTA
Chemical Name or Material trans-Zeatin
Grade HPLC
Show More Show Less

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.