missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin, TRC

Product Code. 30133627
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133627 50 mg 50mg
30133043 25 mg 25mg
30007057 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133627 Supplier Toronto Research Chemicals Supplier No. TRCZ27300050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-39-4
Molecular Formula C10 H13 N5 O
Molecular Weight (g/mol) 219.24
Synonym (2E)-2-Methyl-4-(9H-purin-6-ylamino)-2-buten-1-ol (ACI), 2-Buten-1-ol, 2-methyl-4-(1H-purin-6-ylamino)-, (2E)- (9CI), 2-Buten-1-ol, 2-methyl-4-(1H-purin-6-ylamino)-, (E)- (ZCI), 2-Buten-1-ol, 2-methyl-4-(purin-6-ylamino)-, (E)- (8CI), Zeatin (7CI), (E)-Zeatin, 6-(4-Hydroxy-3-methyl-trans-2-butenylamino)purine, MB 907, N6-(4-Hydroxy-3-methyl-trans-2-butenyl)adenine, trans-6-(4-Hydroxy-3-methylbut-2-enyl)amino purine, trans-Zeatin, Zeatine, ZT, ZTA
IUPAC Name (E)-2-methyl-4-(9H-purin-6-ylamino)but-2-en-1-ol
SMILES C\C(=C/CNc1ncnc2[nH]cnc12)\CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h2,5-6,16H,3-4H2,1H3,(H2,11,12,13,14,15)/b7-2+
Formula Weight 219.112
Percent Purity >95%
Chemical Name or Material trans-Zeatin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.