missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin (synthetic), 97+%

Product Code. 10697231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697231 10 mg Glass bottle 10mg
10480711 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10697231 Supplier Thermo Scientific Acros Supplier No. 264290100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1637-39-4 | C10H13N5O | 219.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-39-4
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.25
MDL Number MFCD00213654
InChI Key UZKQTCBAMSWPJD-FARCUNLSSA-N
Synonym trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
PubChem CID 449093
ChEBI CHEBI:16522
IUPAC Name (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Melting Point 207°C
Color Colorless to Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 10315
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and dimethylformamide
Spectrophotometry 96.0% min. (on dried substance)
Formula Weight 219.25
Percent Purity 97+%
Related Substances 2% max. (HPLC)
Physical Form Liquid
Chemical Name or Material trans-Zeatin (synthetic), 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.