missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin, MedChemExpress

Product Code. 16463797
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463797 50 mg 50mg
16477848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16463797 Supplier Medchem Express Supplier No. HY19700
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress trans-Zeatin is a plant cytokinin, which plays an important role in cell growth, differentiation, and division; trans-Zeatin also inhibits UV-induced MEK/ERK activation.

trans-Zeatin is a plant cytokinin, which plays an important role in cell growth, differentiation, and division

Specifications

Chemical Name or Material trans-Zeatin
Physical Form Solid
CAS 1637-39-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C10H13N5O
Solubility Information DMSO : 100 mg/mL (456.12 mM; Need ultrasonic)
SMILES OC/C(C)=C/CNC1=C2N=CNC2=NC=N1
Molecular Weight (g/mol) 219.24
Formula Weight 219.24
Percent Purity 98.53%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.