missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Vaccenic acid, MedChemExpress
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
trans-Vaccenic acid is a precursor for the synthesis of saturated fatty acid in the rumen and of conjugated linoleic acid (CLA) at the tissue level.
Specifications
Specifications
| Chemical Name or Material | trans-Vaccenic acid |
| Physical Form | Solid |
| CAS | 693-72-1 |
| Color | White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H34O2 |
| Solubility Information | DMSO : 100 mg/mL (354.03 mM; Need ultrasonic) |
| SMILES | CCCCCC/C=C/CCCCCCCCCC(O)=O |
| Molecular Weight (g/mol) | 282.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction